Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osta-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061229.4 | NP_493630.2 | 1 - 2 | 90 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |