Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061267.5 | NP_493668.1 | 8 - 9 | 1009 | 73 | |
GenBank mRNA | X72941.1 | - | 1003 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |