Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027010.5 | NP_001022181.2 | 9 - 10 | 1506 | 76 | |
NM_001313618.3 | NP_001300547.1 | 6 - 7 | 1131 | 76 | ||
NM_001313619.3 | NP_001300548.1 | 1 - 2 | 291 | 76 | ||
NM_001313620.3 | NP_001300549.1 | 6 - 7 | 1146 | 76 | ||
NM_001383772.1 | 11 - 12 | 1634 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |