Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nol-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316867.3 | NP_001303796.1 | 3 - 4 | 800 | 81 | |
NM_061341.6 | NP_493742.1 | 6 - 7 | 1564 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |