Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061349.8 | NP_493750.1 | 8 - 9 | 1178 | 71 | |
GenBank mRNA | AJ252168.1 | - | 8 - 9 | 1090 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |