Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267093.2 | NP_001254022.1 | 3 - 4 | 456 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |