Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047733.5 | NP_001041198.2 | 6 - 7 | 689 | 75 | |
NM_074237.8 | NP_506638.2 | 6 - 7 | 687 | 75 | ||
NM_182356.5 | NP_872156.2 | 7 - 8 | 866 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |