Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061449.7 | NP_493850.2 | 3 - 4 | 1543 | 99 |
Gene Symbol: fbxc-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129156.2 | NP_001122628.1 | 3 - 4 | 1507 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |