Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318283.1 | NP_001305212.1 | 1 - 2 | 522 | 69 | |
NM_061470.6 | NP_493871.2 | 3 - 4 | 920 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |