Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377691.1 | NP_001364697.1 | 1 - 2 | 98 | 83 | |
NM_001377692.2 | NP_001364548.1 | 1 - 2 | 104 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |