Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061602.1 | NP_494003.1 | 2 - 3 | 453 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |