Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322712.3 | NP_001309567.1 | 1 - 2 | 88 | 84 | |
NM_061639.2 | NP_494040.1 | 1 - 2 | 82 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |