Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061683.4 | NP_494084.2 | 4 - 5 | 1457 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |