Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267114.2 | NP_001254043.1 | 2 - 3 | 395 | 132 | |
NM_001267115.3 | NP_001254044.1 | 1 - 2 | 275 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |