Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F07E5.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368571.3 | NP_001355413.1 | 6 - 7 | 2180 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |