Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061787.5 | NP_494188.4 | 4 - 5 | 815 | 135 |
Gene Symbol: srh-195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061785.3 | NP_494186.1 | 1 - 2 | 773 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |