Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061787.5 | NP_494188.4 | 5 - 6 | 933 | 152 |
Gene Symbol: srh-195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061785.3 | NP_494186.1 | 2 - 3 | 891 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |