Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sup-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061932.5 | NP_494333.1 | 7 - 8 | 926 | 70 | |
GenBank mRNA | AF083652.1 | - | 7 - 8 | 870 | 70 | |
AY357729.1 | - | 7 - 8 | 940 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |