Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nep-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061942.3 | NP_494343.2 | 12 - 13 | 1678 | 95 |
Gene Symbol: T05A8.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381354.1 | NP_001368013.1 | 12 - 13 | 1683 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |