Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129178.3 | NP_001122650.1 | 4 - 5 | 586 | 65 | |
NM_061982.7 | NP_494383.2 | 5 - 6 | 685 | 65 | ||
NM_061983.8 | NP_494384.2 | 5 - 6 | 674 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |