Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381362.1 | NP_001368014.1 | 10 - 11 | 3345 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |