Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wht-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062094.4 | NP_494495.3 | 5 - 6 | 630 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |