Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062208.3 | NP_494609.1 | 3 - 4 | 346 | 136 |
Gene Symbol: srx-98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377766.2 | NP_001364762.1 | 3 - 4 | 334 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |