Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062252.3 | NP_494653.2 | 1 - 2 | 92 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |