Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sea-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306544.2 | NP_001293473.1 | 5 - 6 | 2203 | 66 | |
NM_001306546.2 | NP_001293475.1 | 5 - 6 | 2203 | 66 | ||
NM_001313639.3 | NP_001300568.1 | 4 - 5 | 1963 | 66 | ||
NM_001313643.3 | NP_001300572.1 | 3 - 4 | 1477 | 66 | ||
NM_001377772.3 | NP_001364836.1 | 8 - 9 | 2491 | 66 | ||
NM_001377774.1 | NP_001364835.1 | 8 - 9 | 2491 | 66 | ||
NM_001381376.1 | NP_001368518.1 | 4 - 5 | 1963 | 66 | ||
NM_001381378.1 | NP_001368182.1 | 3 - 4 | 1477 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |