Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062310.6 | NP_494711.1 | 4 - 5 | 569 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |