Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062310.6 | NP_494711.1 | 7 - 8 | 851 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |