Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhgf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381382.1 | NP_001368191.1 | 10 - 11 | 2520 | 77 | |
NM_001381384.1 | NP_001367992.1 | 10 - 11 | 2251 | 77 | ||
NM_001383815.1 | 10 - 11 | 2254 | 77 | |||
GenBank mRNA | JQ763434.1 | - | 9 - 10 | 2094 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |