Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srm-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028771.4 | NP_001023942.1 | 5 - 6 | 862 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |