Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182427.1 | NP_872227.1 | 2 - 3 | 446 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |