Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373048.3 | NP_001360011.1 | 10 - 11 | 4276 | 63 | |
NM_072394.7 | NP_504795.1 | 10 - 11 | 4315 | 63 | ||
GenBank mRNA | AY519856.1 | - | 10 - 11 | 4257 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |