Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375186.2 | NP_001362159.1 | 6 - 7 | 855 | 78 | |
NM_001375187.2 | NP_001362160.1 | 6 - 7 | 855 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |