Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270296.3 | NP_001257225.1 | 5 - 6 | 764 | 66 | |
NM_001270297.3 | NP_001257226.1 | 5 - 6 | 696 | 66 | ||
NM_001313376.3 | NP_001300305.1 | 5 - 6 | 696 | 66 | ||
NM_001313377.3 | NP_001300306.1 | 5 - 6 | 696 | 66 | ||
NM_001373559.4 | NP_001359729.1 | 5 - 6 | 696 | 66 | ||
NM_077882.5 | NP_510283.1 | 5 - 6 | 758 | 66 | ||
NM_182454.4 | NP_872254.1 | 5 - 6 | 758 | 66 |
Gene Symbol: Y12A6A.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383667.1 | 5 - 6 | 758 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |