Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lat-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047259.5 | NP_001040724.1 | 10 - 11 | 1485 | 69 | |
GenBank mRNA | AY314772.1 | - | 10 - 11 | 1468 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |