Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383818.1 | 2 - 3 | 194 | 56 | ||
NM_062371.5 | NP_494772.1 | 2 - 3 | 187 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |