Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062384.7 | NP_494785.1 | 1 - 2 | 198 | 126 | |
GenBank mRNA | AF083231.1 | - | 1 - 2 | 136 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |