Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062387.6 | NP_494788.3 | 4 - 5 | 1360 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |