Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062387.6 | NP_494788.3 | 7 - 8 | 3010 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |