Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ulp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318311.3 | NP_001305240.1 | 4 | 333 | 83 | |
NM_062513.5 | NP_494914.1 | 3 - 4 | 333 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |