Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026742.5 | NP_001021913.1 | 10 - 11 | 1638 | 116 | |
NM_062545.8 | NP_494946.2 | 10 - 11 | 1617 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |