Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062564.5 | NP_494965.1 | 1 - 2 | 228 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |