Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062594.3 | NP_494995.2 | 15 - 16 | 4985 | 97 | |
NR_134556.1 | 15 - 16 | 5243 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |