Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356838.3 | NP_001343623.1 | 4 - 5 | 782 | 111 | |
NM_001356841.4 | NP_001343742.1 | 732 | 111 | |||
NM_001383838.1 | 4 - 5 | 599 | 111 | |||
NM_062665.8 | NP_495066.2 | 4 - 5 | 782 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |