Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026798.7 | NP_001021969.1 | 1 - 2 | 127 | 83 | |
NM_001313581.4 | NP_001300510.1 | 1 - 2 | 115 | 83 | ||
NM_001381422.1 | NP_001367521.1 | 1 - 2 | 127 | 83 | ||
NM_001381423.2 | NP_001368512.1 | 1 - 2 | 115 | 83 | ||
NM_001383840.1 | 2 - 3 | 285 | 83 | |||
NM_001383841.1 | 1 - 2 | 114 | 83 | |||
GenBank mRNA | AF000363.1 | - | 1 - 2 | 61 | 83 | |
AY661743.1 | - | 1 - 2 | 61 | 83 | ||
AY661745.1 | - | 1 - 2 | 61 | 83 | ||
AY661746.1 | - | 1 - 2 | 61 | 83 | ||
AY661747.1 | - | 1 - 2 | 61 | 83 |
Gene Symbol: C17G10.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313579.3 | NP_001300508.1 | 2 | 115 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |