Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026798.7 | NP_001021969.1 | 3 - 4 | 420 | 87 | |
NM_001313581.4 | NP_001300510.1 | 3 - 4 | 408 | 87 | ||
NM_001381422.1 | NP_001367521.1 | 3 - 4 | 420 | 87 | ||
NM_001381423.2 | NP_001368512.1 | 3 - 4 | 408 | 87 | ||
NM_001383840.1 | 4 - 5 | 578 | 87 | |||
NM_001383841.1 | 3 - 4 | 407 | 87 | |||
GenBank mRNA | AF000363.1 | - | 3 - 4 | 354 | 87 | |
AY661743.1 | - | 3 - 4 | 354 | 87 | ||
AY661745.1 | - | 3 - 4 | 354 | 87 | ||
AY661746.1 | - | 3 - 4 | 354 | 87 | ||
AY661747.1 | - | 3 - 4 | 354 | 87 |
Gene Symbol: C17G10.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313579.3 | NP_001300508.1 | 4 | 408 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |