Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381422.1 | NP_001367521.1 | 4 - 5 | 1691 | 68 | |
NM_001383840.1 | 5 - 6 | 1849 | 68 | |||
NM_001383841.1 | 4 - 5 | 1678 | 68 | |||
GenBank mRNA | AF000363.1 | - | 4 - 5 | 1625 | 68 | |
AY661743.1 | - | 4 - 5 | 1625 | 68 | ||
AY661745.1 | - | 4 - 5 | 1625 | 68 | ||
AY661746.1 | - | 4 - 5 | 1625 | 68 |
Gene Symbol: C17G10.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313579.3 | NP_001300508.1 | 1679 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |