Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zyg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062702.6 | NP_495103.1 | 3 - 4 | 966 | 82 | |
GenBank mRNA | AF285179.1 | - | 3 - 4 | 988 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |