Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062705.10 | NP_495106.1 | 10 - 11 | 1824 | 84 | |
GenBank mRNA | AJ745072.1 | - | 6 - 7 | 978 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |