Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026802.3 | NP_001021973.1 | 6 - 7 | 726 | 130 | |
NM_001381435.2 | NP_001367527.1 | 5 - 6 | 822 | 130 | ||
GenBank mRNA | AB112928.1 | - | 4 - 5 | 579 | 130 | |
U68265.1 | - | 4 - 5 | 601 | 130 | ||
U68266.1 | - | 5 - 6 | 663 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |