Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062782.4 | NP_495183.1 | 1 - 2 | 206 | 98 | |
GenBank mRNA | AY048153.1 | - | 1 - 2 | 206 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |